Masterflex® Turbine Flowmeter, NSF-Approved, 29 to 343 Hz Frequency Range; 0.4 to 4 GPM
Supplier: Avantor Fluid Handling
VWR Item#MFLA98516-92
Your Organization's Price:
Retrieving
Designed for low-viscosity flow measurement in water and beverage applications.
  • NSF Standard 61 listed—ideal for use with water sampling and processing applications
  • Designed with 3/8” NPT(M) process connections
  • Suitable for use with 1.8 to 16 cSt liquids
  • Features an accuracy of ±2% of reading
Designed for low-flow OEM and pilot applications, these turbine flowmeters are highly accurate and repeatable and feature a Hall effect sensor for superior accuracy. The sensor’s standard power and output specifications make it easy to retrofit existing controllers and output to a PLC, recorder, or panel meter. The 316 stainless steel shaft coupled with Delrin® acetal bearings allows for accurate measurements during quick dispensing cycles. These low-cost units meet the requirements of NSF Standard 61, making them ideal for water or beverage dispensing applications or any application with water-based liquids.