GF Signet 161-323-551 Piping System Metering Ball Valve, PVC, 3/8" NPT(F)
Supplier: Avantor Fluid Handling
VWR Item#MFLB01360-40
Your Organization's Price:
Retrieving
Precise control over 180° range.
  • Choose between PVC or polypropylene (PP) construction
Ball valves give you precise control over the entire 180° range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5° increments.