Single open end serial nylon cartridge, 0.45/0.20 µm
Supplier: Avantor Fluid Handling
VWR Item#MFLB06479-18
Your Organization's Price:
Retrieving
A collection to fit a wide range of critical applications.
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
These SOE cartridges have a 222 O-ring configuration to create a tight seal and prevent influent bypass. They are autoclavable to 250 °F (121 °C) and in-line sterilizable to 257 °F (125 °C) for up to 30-minute cycles; tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180 °F (82 °C) at 10 psid. Use a SOE cartridge-sump adapter kit (sold separately) to secure cartridges within housings.<br /><br />The cartridges comply with US CFR Title 21 guidelines for repeated food contact, comply with USP Class VI-121 °C Plastics guidelines and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL and typically exhibit low levels of non-volatile residues.
Multi-Layer Nylon Cartridges Two-pleated nylon<br />membranes with the top layer acting as a prefilter<br />to increase cartridge life and efficiency.