Gems Standard Level Switch, Polypropylene, 0.90 Minimum Specific Gravity, 1/8" NPT
Supplier: Avantor Fluid Handling
VWR Item#MFLB07187-10
Your Organization's Price:
Retrieving
Install from the top of your tank—attach your own conduit to protect wiring.
  • Get longer life and more durability with hermetically sealed switches
All models accept 120/240 VAC and operate as NO or NC switches except where noted. Switches include 24" leads (except 07187-12 which has 72" leads). If your level control system exceeds the current rating in the table, use a controller with your switch.