Flowline LU10-1305 Ultrasonic Level Switch-PP, Short, 3/4" NPT
Supplier: Avantor Fluid Handling
VWR Item#MFLB23002-14
Your Organization's Price:
Retrieving
Compatible with Smart Trak and Switch-Pak installation fittings.
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.