Flowline LI55-1001 Advanced Level Control Meter, No Outputs; 24 VDC
Supplier: Avantor Fluid Handling
VWR Item#MFLB23002-18
Your Organization's Price:
Retrieving
Easy configuration via push button display module.
  • Bright 6-digit LED main display and secondary unit display with relay status indicators
  • 1/8 DIN enclosure with NEMA 4X faceplate and shallow depth case
  • 32-point linearization function for volumetric tank measurement
  • Non-volatile memory with security password protected lock out
  • Isolated 4-20 mA repeater output with 40 mA internal power supply
The general purpose meter displays engineering units with one 4-20 mA or 0-10 VDC transmitter input channel and is offered in three configurations with optional 2 or 4 relays, 4 or 8 set points and an isolated 4-20 mA repeater. Each relay can be configured on a single set point or latched on two set points for automatic fill or empty control including duplexing. For field mount installations, <br />order the single or two meter NEMA box.