Fitting, Stainless Steel, Straight, Compression Union, 5/16" OD
Supplier: Avantor Fluid Handling
VWR Item#MFLB31406-50
Your Organization's Price:
Retrieving
Get a secure seal at higher pressures.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).