GF Signet 3-2552-21-A-12 Magmeter Flow Sensor, 9.3" Depth, 25' Cable, 4-20 mA
Supplier: Avantor Fluid Handling
VWR Item#MFLB32486-45
Your Organization's Price:
Retrieving
Optimize performance of your GF+ Signet Transmitter/Controller.
  • Choose from a wide range of replacement parts and accessories
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
Should be used in conjunction with a 56560-12 or 56560-14 9900 Series Transmitter, and 56560-15 relay module in order to create a Batch Controller System.