Masterflex® Sterile Forceps, stainless steel, 4.5" long; 25/pk
Supplier: Avantor Fluid Handling
VWR Item#MFLB75955-13
Your Organization's Price:
Retrieving
No more cross-contamination.
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.