Masterflex® Printed Microscope Slide, ten rectangular sections; 144/pk
Supplier: Avantor Fluid Handling
VWR Item#MFLB75955-31
Your Organization's Price:
Retrieving
Optimize sample testing and viewing.
  • Screen-printed section design keeps samples contained
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.