Masterflex® PTFE Check valve, 1/2" Orifice, 1/2" Female NPT
Supplier: Avantor Fluid Handling
VWR Item#MFLB98515-18
Your Organization's Price:
Retrieving
Use with caustic and high-purity liquids in many applications.
  • Constructed of durable PTFE
Designed for use with corrosive fluids of varying viscosities in chemical, microelectronics, and petroleum industries.