Masterflex® L/S® Variable-Speed Compact Drive, 35 to 200 rpm; 115/230 VAC
Supplier: Avantor Fluid Handling
VWR Item#MFLA77200-20-IL
Your Organization's Price:
Retrieving
Stackable, compact pump drive delivers versatile performance in limited space.
  • Flow rate 2.1 to 560 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Stackable, compact drive is perfect for small work areas
  • Constructed with a painted steel housing with IP22 rating
  • No difficult programming—single switch for forward/off/reverse and analog controller
  • Pump in either direction—also allows purging before or after pumping
  • Remote control of start/stop via contact closure
  • Runs on 12 VDC power—order power supply 77200-07 separately
These compact drives are a perfect when space is limited, as they fit just about anywhere. The variable-speed design makes them ideal for when you need to pump fluid at a higher rate. Drives accept one Easy-Load® or standard pump head. Use drives with L/S® Precision tubing—accepts sizes L/S 13, L/S 14, L/S 16, L/S 25, and L/S 17. Only silicone and C-Flex® tubing formulations are recommended for use with L/S 25 and L/S 17 sizes.
<b>Delivery information: </b>Includes dual voltage benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.