Masterflex® Optical Sensor Controller for Single-Point Detection And Control
Supplier: Avantor Fluid Handling
VWR Item#MFLB32772-50
Your Organization's Price:
Retrieving
Converts standard 110 VAC line current to 10 to 28 VDC input power.
  • Easily covert VAC to VDC
Controller converts standard 110 VAC line current to the 10 to 28 VDC input power required for optical sensor operation, and provides an SPDT, 5 A relay output for direct control of moderate loads. Input voltage: 120 VAC, 50/60Hz. Max current draw: 70 mA at 120 VAC. Operating temperature: –13 to 158 °F (–25 to 70 °C). Electrical connections: eight ¼" male spade terminals. Dimensions: 3"L×23-1/32"W×4-15/16"H.