Hubtron Transfer Tubing, Hubtron™ Black Advance Fluoroelastomer, 1/16" ID x 3/16" OD; 25 Ft
Supplier: Avantor Fluid Handling
VWR Item#MFLB95256-00
Your Organization's Price:
Retrieving
Ideal for corrosive chemical transfer.
  • PTFE tubing has 1/16" inner diameter
<B>Applications:</B> Corrosive chemical transfer, electronics applications, ink and printing, and general manufacturing applications requiring UV protection.|<B>Characteristics:</B> Opaque black with UV block, very low gas permeability, highly chemically compatible. Inner fluoroplastic liner, outer fluoroelastomer cover. Excellent chemical resistance to acids, alkalis, alcohols, and esters.<br /><B>Certification:</B> None.<br /><B>Temperature:</B> 14 to 302 °F (–5 to 150 °C)<br /><B>Sterilization:</B> Not recommended.