Fitting, Polypropylene, Y-Connector, Hose Barb Union, 14 mm ID; 100/Pk
Supplier: Avantor Fluid Handling
VWR Item#MFLA06295-60BLK
Your Organization's Price:
Retrieving
No tools needed for assembly.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 14 mm ID; 100/PK
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.