(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium

38596.03 38596.06
ALFA38596.03EA 129 SGD
ALFA38596.03 ALFA38596.06
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium

Purity: 98% CAS No.: 22441-13-0 Molecular Formula: C11H19LiO2 Formula Weight: 190.24 Physical Form: Powder Melting Point: 127° Boiling Point: 180°/0.05mm MDL No.: MFCD00142754

Formula: C₁₁H₁₉LiO₂
MW: 190,24 g/mol
Boiling Pt: 180 °C (0,05 mmHg)
Melting Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00142754
CAS Number: 22441-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR