Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥99.9%

Supplier: Thermo Scientific
Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), Bi(TMHD)3

41202.03 41202.06 41202.MD
ALFA41202.03EA 127 SGD
ALFA41202.03 ALFA41202.06 ALFA41202.MD
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥99.9%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)

Purity: 99.9% CAS No.: 142617-53-6 Molecular Formula: C33H57BiO6 Formula Weight: 758.79 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: White to brown Melting Point: 114-116° Boiling Point: 150°/0.05mm MDL No.: MFCD00064763

Formula: C₃₃H₅₇BiO₆
MW: 758,782 g/mol
Boiling Pt: 150 °C (0,05 mmHg)
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00064763
CAS Number: 142617-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR