Fmoc-D-phenylalanine ≥98%
Supplier: Thermo Scientific
Synonyms:
Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine
B21689.03
B21689.06
B21689.14
ALFAB21689.03EA
67.9
SGD
ALFAB21689.03
ALFAB21689.06
ALFAB21689.14
Fmoc-D-phenylalanine ≥98%
Fmoc-D-phenylalanine
Purity: 98%
CAS No.: 86123-10-6
Molecular Formula: C24H21NO4
Formula Weight: 387.44
Storage Temperature: 2°C to 8°C
Melting Point: 181-185°
Optical Rotation: +40° (c=1 in DMF)
MDL No.: MFCD00062955
Formula:
C₂₄H₂₁NO₄ MW: 387,4 g/mol Melting Pt: 181…185 °C |
MDL Number:
MFCD00062955 CAS Number: 86123-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...