Chlorobis(2-methoxyphenyl)phosphine ≥98%

Supplier: Thermo Scientific
Danger

H25894.06 H25894.03
ALFAH25894.06EA 0 SGD
ALFAH25894.06 ALFAH25894.03
Chlorobis(2-methoxyphenyl)phosphine ≥98%
Chlorobis(2-methoxyphenyl)phosphine

Purity: 98+% CAS No.: 263369-88-6 Molecular Formula: C14H14ClO2P Formula Weight: 280.69 Melting Point: 111-114° UN No.: 3261 MDL No.: MFCD08064037

Formula: C₁₄H₁₄ClO₂P
MW: 280,69 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD08064037
CAS Number: 263369-88-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR