Chlorobis(4-methoxyphenyl)phosphine ≥98%
Supplier: Thermo Scientific
|
Danger
|
H25887.06
H25887.03
ALFAH25887.06EA
897
SGD
ALFAH25887.06
ALFAH25887.03
Chlorobis(4-methoxyphenyl)phosphine ≥98%
Chlorobis(4-methoxyphenyl)phosphine
Purity: 98+%
CAS No.: 13685-30-8
Molecular Formula: C14H14ClO2P
Formula Weight: 280.69
Odour: Irritating
Melting Point: 61-64°
Boiling Point: 140°/1.5mm
UN No.: 3261
MDL No.: MFCD04972301
Formula:
C₁₄H₁₄ClO₂P MW: 280,69 g/mol Boiling Pt: 140 °C (1,5 mmHg) Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972301 CAS Number: 13685-30-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...