3-Amino-1H-pyrazole-4-carboxylic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Amino-4-pyrazolecarboxylic acid, 3-Aminopyrazole-4-carboxylic acid, 3-Amino-1H-pyrazole-4-carboxylic acid, NSC 89246

B23751.03 B23751.06
ALFAB23751.03EA 0 SGD
ALFAB23751.03 ALFAB23751.06
3-Amino-1H-pyrazole-4-carboxylic acid 95%
3-Amino-1H-pyrazole-4-carboxylic acid

Purity: 95% CAS No.: 41680-34-6 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Storage Temperature: 2°C to 8°C Melting Point: ca 125° dec. MDL No.: MFCD00005239

Formula: C₄H₅N₃O₂
MW: 127,10 g/mol
Melting Pt: ∼ 125 °C (dec.)
MDL Number: MFCD00005239
CAS Number: 41680-34-6
EINECS: 255-493-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR