2-Hydrazinobenzothiazole ≥97%

Supplier: Thermo Fisher Scientific
Danger

A18006.09 A18006.18 A18006.30
ALFAA18006.09EA 0 SGD
ALFAA18006.09 ALFAA18006.18 ALFAA18006.30
2-Hydrazinobenzothiazole ≥97%
2-Hydrazinobenzothiazole

Purity: 97% CAS No.: 615-21-4 Molecular Formula: C7H7N3S Formula Weight: 165.21 Melting Point: 198-202° MDL No.: MFCD00041849

Formula: C₇H₇N₃S
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00041849
CAS Number: 615-21-4
EINECS: 210-416-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR