N,N-Dimethylcyanoacetamide 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Cyano-N,N-dimethylacetamide

A10981.06 A10981.14
ALFAA10981.06EA 0 SGD
ALFAA10981.06 ALFAA10981.14
N,N-Dimethylcyanoacetamide 98
N,N-Dimethylcyanoacetamide

Purity: 98% CAS No.: 7391-40-4 Molecular Formula: NCCH2CON(CH3)2 Formula Weight: 112.13 Melting Point: 59-66° Beilstein Reference: 773950

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 59…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00051623
CAS Number: 7391-40-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR