3-Amino-3-(4-chlorophenyl)propionic acid 97%

Supplier: Thermo Scientific

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(p-chlorophenyl)propionic acid

L19798.03 L19798.06 L19798.MD
ALFAL19798.03EA 0 SGD
ALFAL19798.03 ALFAL19798.06 ALFAL19798.MD
3-Amino-3-(4-chlorophenyl)propionic acid 97%
3-Amino-3-(4-chlorophenyl)propionic acid

Purity: 97% CAS No.: 19947-39-8 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Storage Temperature: 2°C to 8°C Melting Point: 214-216° dec. Beilstein Reference: 2365902 MDL No.: MFCD00126370

Formula: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Melting Pt: 214…216 °C (dec.)
MDL Number: MFCD00126370
CAS Number: 19947-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR