3-Amino-3-(4-fluorophenyl)propionic acid 96%
Supplier: Thermo Scientific
|
Warning
|
L19799.03
L19799.MD
ALFAL19799.03EA
0
SGD
ALFAL19799.03
ALFAL19799.MD
3-Amino-3-(4-fluorophenyl)propionic acid 96%
3-Amino-3-(4-fluorophenyl)propionic acid
Purity: 96%
CAS No.: 325-89-3
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Storage Temperature: 2°C to 8°C
Melting Point: 214° dec.
Beilstein Reference: 3258498
MDL No.: MFCD00181811
Formula:
C₉H₁₀FNO₂ MW: 183,18 g/mol Melting Pt: 214 °C (dec.) |
MDL Number:
MFCD00181811 CAS Number: 325-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...