Potassium-4-butoxyphenyltrifluoroborate 96%

Supplier: Thermo Scientific
Warning

H53092.MD H53092.03
ALFAH53092.MDEA 78.1 SGD
ALFAH53092.MD ALFAH53092.03
Potassium-4-butoxyphenyltrifluoroborate 96%
Potassium 4-butoxyphenyltrifluoroborate

Purity: 96% CAS No.: 850623-61-9 Molecular Formula: C10H13BF3KO Formula Weight: 256.12 Melting Point: 300° MDL No.: MFCD04115755

Formula: C₁₀H₁₃BF₃KO
MW: 256,12 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115755
CAS Number: 850623-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR