2-Amino-4-(2,4-difluorophenyl)thiazole 97%

Supplier: Thermo Scientific
Danger

H31948.ME H31948.04
ALFAH31948.MEEA 0 SGD
ALFAH31948.ME ALFAH31948.04
2-Amino-4-(2,4-difluorophenyl)thiazole 97%
2-Amino-4-(2,4-difluorophenyl)thiazole

Purity: 97% CAS No.: 105512-80-9 Molecular Formula: C9H6F2N2S Formula Weight: 212.22 MDL No.: MFCD00472891

Formula: C₉H₆F₂N₂S
MW: 212,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00472891
CAS Number: 105512-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR