3-(Hydroxymethyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

L15193.03 L15193.06
ALFAL15193.03EA 140 SGD
ALFAL15193.03 ALFAL15193.06
3-(Hydroxymethyl)phenylboronic acid ≥98%
3-(Hydroxymethyl)phenylboronic acid

Purity: 98% CAS No.: 87199-15-3 Molecular Formula: C7H9BO3 Formula Weight: 151.96 Melting Point: 93-98° Beilstein Reference: 8832019

Formula: C₇H₉BO₃
MW: 151,96 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01317846
CAS Number: 87199-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR