2-Chloro-4-iodonicotinic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Chloro-4-iodopyridine-3-carboxylic acid
H50049.03
H50049.06
ALFAH50049.03EA
0
SGD
ALFAH50049.03
ALFAH50049.06
2-Chloro-4-iodonicotinic acid ≥98%
2-Chloro-4-iodonicotinic acid
Purity: 98%
CAS No.: 544671-78-5
Molecular Formula: C6H3ClINO2
Formula Weight: 283.45
Melting Point: 162-167°
MDL No.: MFCD05663520
Formula:
C₆H₃ClINO₂ MW: 283,45 g/mol Melting Pt: 162…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663520 CAS Number: 544671-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...