Chlorobis(2-methylphenyl)phosphine 98%, rods

Supplier: Thermo Scientific
Danger

Synonyms: Di-o-tolylchlorophosphine, Chlorodi(o-tolyl)phosphine

H25890.06 H25890.03
ALFAH25890.06EA 621 SGD
ALFAH25890.06 ALFAH25890.03
Chlorobis(2-methylphenyl)phosphine 98%, rods
Chlorobis(2-methylphenyl)phosphine

Purity: 98% CAS No.: 36042-94-1 Molecular Formula: C14H14ClP Formula Weight: 248.69 Melting Point: 57° Boiling Point: 174-178°/3mm UN No.: 1760 MDL No.: MFCD04038733

Formula: C₁₄H₁₄ClP
MW: 248,688 g/mol
Boiling Pt: 174…178 °C (3 mmHg)
Melting Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD04038733
CAS Number: 36042-94-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR