4-Bromo-trans-stilbene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: trans-4-Bromostilbene

H30189.03 H30189.06
ALFAH30189.03EA 0 SGD
ALFAH30189.03 ALFAH30189.06
4-Bromo-trans-stilbene ≥98%
4-Bromo-trans-stilbene

Purity: 98% CAS No.: 13041-70-8 Molecular Formula: C14H11Br Formula Weight: 259.15 Melting Point: 138-140° MDL No.: MFCD00155020

Formula: C₁₄H₁₁Br
MW: 259.15 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00155020
CAS Number: 13041-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR