2-(Bromomethyl)benzothiazole 98

Supplier: Thermo Fisher Scientific
Danger

H26120.MD H26120.03 H26120.06
ALFAH26120.MDEA 0 SGD
ALFAH26120.MD ALFAH26120.03 ALFAH26120.06
2-(Bromomethyl)benzothiazole 98
2-(Bromomethyl)benzothiazole

Purity: 98+% CAS No.: 106086-78-6 Molecular Formula: C8H6BrNS Formula Weight: 228.11 Storage Temperature: 2°C to 8°C Melting Point: 47-49° UN No.: 3261 MDL No.: MFCD00461195

Formula: C₈H₆BrNS
MW: 228.11 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD00461195
CAS Number: 106086-78-6
EINECS: None
UN: 3261
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR