3-Amino-5-tert-butyl-1H-pyrazole 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Amino-5-tert-butylpyrazole
A12750.06
A12750.03
A12750.14
ALFAA12750.06EA
474
SGD
ALFAA12750.06
ALFAA12750.03
ALFAA12750.14
3-Amino-5-tert-butyl-1H-pyrazole 98%
3-Amino-5-tert-butyl-1H-pyrazole
Purity: 98%
CAS No.: 82560-12-1
Molecular Formula: C7H13N3
Formula Weight: 139.20
Melting Point: 75-79°
MDL No.: MFCD00067987
Formula:
C₇H₁₃N₃ MW: 139,20 g/mol Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067987 CAS Number: 82560-12-1 EINECS: 450-360-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...