4-tert-Butyl-3-nitrophenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-tert-Butyl-3-nitrobenzeneboronic acid

H52883.MD H52883.03
ALFAH52883.MDEA 167 SGD
ALFAH52883.MD ALFAH52883.03
4-tert-Butyl-3-nitrophenylboronic acid ≥98%
4-tert-Butyl-3-nitrophenylboronic acid

Purity: 98% CAS No.: 871329-54-3 Molecular Formula: C10H14BNO4 Formula Weight: 223.04 Melting Point: 52-54° MDL No.: MFCD07363743

Formula: C₁₀H₁₄BNO₄
MW: 223,04 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07363743
CAS Number: 871329-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR