2-(1-Piperidinyl)phenol ≥98%
Supplier: Thermo Scientific
|
Warning
|
L07630.03
L07630.06
ALFAL07630.03EA
118
SGD
ALFAL07630.03
ALFAL07630.06
2-(1-Piperidinyl)phenol ≥98%
2-(1-Piperidinyl)phenol
Purity: 98%
CAS No.: 65195-20-2
Molecular Formula: C11H15NO
Formula Weight: 177.24
Melting Point: 73-76°
Beilstein Reference: 151777
Formula:
C₁₁H₁₅NO MW: 177,24 g/mol Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051687 CAS Number: 65195-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...