Furfuryl methyl sulphide ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-((Methylthio)methyl)furan
B21691.06
B21691.14
B21691.22
ALFAB21691.06EA
0
SGD
ALFAB21691.06
ALFAB21691.14
ALFAB21691.22
Furfuryl methyl sulphide ≥98%
Furfuryl methyl sulphide
Purity: 98%
CAS No.: 1438-91-1
Molecular Formula: C6H8OS
Formula Weight: 128.19
Flash Point: 63°(145°F)
Refractive Index: 1.5240
Boiling Point: 170-173°
Beilstein Reference: 107109
MDL No.: MFCD00009604
Formula:
C₆H₈OS MW: 128,19 g/mol Boiling Pt: 170…173 °C Density: 1,070 Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009604 CAS Number: 1438-91-1 EINECS: 215-874-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...