4-Indoleboronic acid 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Indolylboronic acid, (1H-Indol-4-yl)boronic acid
H53037.MD
H53037.03
ALFAH53037.MDEA
244
SGD
ALFAH53037.MD
ALFAH53037.03
4-Indoleboronic acid 95%
4-Indoleboronic acid
Purity: 95%
CAS No.: 220465-43-0
Molecular Formula: C8H8BNO2
Formula Weight: 160.97
Storage Temperature: -30°C to -10°C
Formula:
C₈H₈BNO₂ MW: 160,97 g/mol |
MDL Number:
MFCD03095175 CAS Number: 220465-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...