3-(2-Carboxyvinyl)phenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(3-Boronophenyl)acrylic acid, 3-(2-Carboxyvinyl)benzeneboronic acid
L16369.03
L16369.MD
ALFAL16369.03EA
0
SGD
ALFAL16369.03
ALFAL16369.MD
3-(2-Carboxyvinyl)phenylboronic acid ≥97%
3-(2-Carboxyvinyl)phenylboronic acid
Purity: 97%
CAS No.: 216144-91-1
Molecular Formula: C9H9BO4
Formula Weight: 191.98
Melting Point: ca 218°
MDL No.: MFCD01075738
Formula:
C₉H₉BO₄ MW: 191,98 g/mol Melting Pt: ∼ 218 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075738 CAS Number: 216144-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...