4-(Dimethylcarbamoyl)-3-fluorophenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid

H53117.MD H53117.03
ALFAH53117.MDEA 0 SGD
ALFAH53117.MD ALFAH53117.03
4-(Dimethylcarbamoyl)-3-fluorophenylboronic acid ≥98%
4-(Dimethylcarbamoyl)-3-fluorophenylboronic acid

Purity: 98% CAS No.: 874289-13-1 Molecular Formula: C9H11BFNO3 Formula Weight: 211.00 Melting Point: 147-149° MDL No.: MFCD08436060

Formula: C₉H₁₁BFNO₃
MW: 211,00 g/mol
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD08436060
CAS Number: 874289-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR