2-Fluoro-5-(methylcarbamoyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid, 2-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

H53178.MD H53178.03
ALFAH53178.MDEA 0 SGD
ALFAH53178.MD ALFAH53178.03
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid ≥98%
2-Fluoro-5-(methylcarbamoyl)phenylboronic acid

Purity: 98% CAS No.: 874289-40-4 Molecular Formula: C8H9BFNO3 Formula Weight: 196.97 Melting Point: 174-176° MDL No.: MFCD08436028

Formula: C₈H₉BFNO₃
MW: 196,97 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD08436028
CAS Number: 874289-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR