3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

H53084.MD H53084.03
ALFAH53084.MDEA 0 SGD
ALFAH53084.MD ALFAH53084.03
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 95%
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid

Purity: 95% CAS No.: 871329-62-3 Molecular Formula: C8H8BFO4 Formula Weight: 197.96 Melting Point: 138-142° MDL No.: MFCD07363749

Formula: C₈H₈BFO₄
MW: 197,96 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD07363749
CAS Number: 871329-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR