4-(Methylsulphonyl)phenol ≥97%
Supplier: Thermo Scientific
|
Warning
|
H32935.03
H32935.09
ALFAH32935.03EA
0
SGD
ALFAH32935.03
ALFAH32935.09
4-(Methylsulphonyl)phenol ≥97%
4-(Methylsulphonyl)phenol
Purity: 97%
CAS No.: 14763-60-1
Molecular Formula: C7H8O3S
Formula Weight: 172.20
Melting Point: 90-95°
MDL No.: MFCD00047806
Formula:
C₇H₈O₃S Melting Pt: 90…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047806 CAS Number: 14763-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...