3-(2-Carboxyethyl)phenylboronic acid 96%
Supplier: Thermo Scientific
|
Warning
|
H52977.MD
H52977.03
ALFAH52977.MDEA
0
SGD
ALFAH52977.MD
ALFAH52977.03
3-(2-Carboxyethyl)phenylboronic acid 96%
3-(2-Carboxyethyl)phenylboronic acid
Purity: 96%
CAS No.: 693803-17-7
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 166-169°
MDL No.: MFCD04115649
Formula:
C₉H₁₁BO₄ MW: 193,99 g/mol Melting Pt: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115649 CAS Number: 693803-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...