4-Bromo-2-fluorobenzylamine hydrochloride 97%

Supplier: Thermo Scientific
Warning

Synonyms: (4-Bromo-2-fluorophenyl)methanamine hydrochloride

A15654.06 A15654.14
ALFAA15654.06EA 0 SGD
ALFAA15654.06 ALFAA15654.14
4-Bromo-2-fluorobenzylamine hydrochloride 97%
4-Bromo-2-fluorobenzylamine hydrochloride

Purity: 98% CAS No.: 147181-08-6 Molecular Formula: C7H7BrFN.HCl Formula Weight: 240.50 Melting Point: 254-257° Beilstein Reference: 6938847

Formula: C₇H₇BrFN·HCl
MW: 240,50 g/mol
Melting Pt: 254…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00143266
CAS Number: 147181-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR