3,4-Dihydroxybenzylamine hydrobromide ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(Aminomethyl)benzene-1,2-diol hydrobromide

B21833.03 B21833.06 B21833.14
ALFAB21833.03EA 0 SGD
ALFAB21833.03 ALFAB21833.06 ALFAB21833.14
3,4-Dihydroxybenzylamine hydrobromide ≥98%
3,4-Dihydroxybenzylamine hydrobromide

Purity: 98% CAS No.: 16290-26-9 Molecular Formula: C7H9NO2.HBr Formula Weight: 220.07 Melting Point: 196-200° Beilstein Reference: 4002646

Formula: C₇H₁₀BrNO₂
MW: 220,07 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00012859
CAS Number: 16290-26-9
EINECS: 240-382-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR