4-Chloro-2,6-xylidine hydrochloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Chloro-2,6-dimethylaniline hydrochloride

A19362.09 A19362.18
ALFAA19362.09EA 0 SGD
ALFAA19362.09 ALFAA19362.18
4-Chloro-2,6-xylidine hydrochloride ≥98%
4-Chloro-2,6-xylidine hydrochloride

Purity: 98% CAS No.: 5769-32-4 Molecular Formula: C8H10ClN.HCl Formula Weight: 192.09 MDL No.: MFCD00671703

Formula: C₈H₁₁Cl₂N
MW: 192,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671703
CAS Number: 5769-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR