3-Biphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

L17552.03 L17552.06
ALFAL17552.03EA 0 SGD
ALFAL17552.03 ALFAL17552.06
3-Biphenylboronic acid ≥98%
3-Biphenylboronic acid

Purity: 98% CAS No.: 5122-95-2 Molecular Formula: C12H11BO2 Formula Weight: 198.03 Odour: Odourless Melting Point: 206-211° Beilstein Reference: 2836311 MDL No.: MFCD01318102

Formula: C₁₂H₁₁BO₂
MW: 198,03 g/mol
Melting Pt: 206…211 °C
Storage Temperature: Ambient
MDL Number: MFCD01318102
CAS Number: 5122-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR