4-(Heptyloxy)phenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid
H53066.03
H53066.06
ALFAH53066.03EA
64.5
SGD
ALFAH53066.03
ALFAH53066.06
4-(Heptyloxy)phenylboronic acid ≥97%
4-(Heptyloxy)phenylboronic acid
Purity: 97%
CAS No.: 136370-19-9
Molecular Formula: C13H21BO3
Formula Weight: 236.12
MDL No.: MFCD04039169
Formula:
C₁₃H₂₁BO₃ MW: 236,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039169 CAS Number: 136370-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...