5-Fluoro-2-methylphenylboronic acid ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

L19819.03 L19819.06
ALFAL19819.03EA 0 SGD
ALFAL19819.03 ALFAL19819.06
5-Fluoro-2-methylphenylboronic acid ≥99%
5-Fluoro-2-methylphenylboronic acid

Purity: 99% CAS No.: 163517-62-2 Molecular Formula: C7H8BFO2 Formula Weight: 153.95 Melting Point: 148-149° MDL No.: MFCD03095047

Formula: C₇H₈BFO₂
MW: 153,95 g/mol
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03095047
CAS Number: 163517-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR