4-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

H52483.03 H52483.06
ALFAH52483.03EA 143 SGD
ALFAH52483.03 ALFAH52483.06
4-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥98%
4-Fluoro-3-(trifluoromethyl)phenylboronic acid

Purity: 98% CAS No.: 182344-23-6 Molecular Formula: C7H5BF4O2 Formula Weight: 207.92 Melting Point: 170-174° MDL No.: MFCD07363787

Formula: C₇H₅BF₄O₂
MW: 207,92 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD07363787
CAS Number: 182344-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR